Learn More
2-Bromo-4-nitrophenol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-4-nitrophenol |
| CAS | 5847-59-6 |
| Molecular Formula | C6H4BrNO3 |
| Quantity | 250 mg |
| Synonym | NSC 212120 |
| InChI Formula | InChI=1S/C6H4BrNO3/c7-5-3-4(8(10)11)1-2-6(5)9/h1-3,9H |
| SMILES | Oc1ccc(cc1Br)[N+](=O)[O-] |
| IUPAC Name | 2-bromo-4-nitrophenol |
| Molecular Weight (g/mol) | 218.01 |
| Formula Weight | 216.9375 |