missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trifluoromethyl)phenol, 98%, Thermo Scientific Chemicals

Catalog Number p-7055431
38.13 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
402-05-1
C7H4BrF3O
241.007
MFCD09261190
INHVNZLKNPJCJD-UHFFFAOYSA-N
2-bromo-5-trifluoromethyl phenol, phenol, 2-bromo-5-trifluoromethyl, acmc-209jbu, phenol,2-bromo-5-trifluoromethyl, 3-bromo-5-hydroxybenzotrifluoride, 4-bromo-3-hydroxybenzotrifluoride
21914470
2-bromo-5-(trifluoromethyl)phenol
C1=CC(=C(C=C1C(F)(F)F)O)Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
402-05-1
C7H4BrF3O
241.007
MFCD09261190
INHVNZLKNPJCJD-UHFFFAOYSA-N
2-bromo-5-trifluoromethyl phenol, phenol, 2-bromo-5-trifluoromethyl, acmc-209jbu, phenol,2-bromo-5-trifluoromethyl, 3-bromo-5-hydroxybenzotrifluoride, 4-bromo-3-hydroxybenzotrifluoride
21914470
2-bromo-5-(trifluoromethyl)phenol
C1=CC(=C(C=C1C(F)(F)F)O)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Quantity 5 g
UN Number UN3265
Formula Weight 241.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)phenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'