missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-L-phenylalanine, 95%

Brand:  Alfa Aesar™ H63141.MD

Additional Details : CAS : 42538-40-9 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15439668

  • € 27.70 / 250mg


Estimated Shipment date
from Supplier 06-07-2020
Add to basket

Specifications

Chemical Identifiers

42538-40-9
244.088
JFVLNTLXEZDFHW-QMMMGPOBSA-N
193338
C1=CC=C(C(=C1)CC(C(=O)O)N)Br
C9H10BrNO2
MFCD03092958
l-2-bromophenylalanine, 2-bromo-l-phenylalanine, s-2-amino-3-2-bromophenyl propanoic acid, 2-bromophenylalanine, 2s-2-amino-3-2-bromophenyl propanoic acid, h-phe 2-br-oh, o-bromo-l-phenylalanine, l-2-br-phe-oh, s-2-bromophenylalanine, l-2-bromophe
(2S)-2-amino-3-(2-bromophenyl)propanoic acid

Specifications

2-Bromo-L-phenylalanine
C9H10BrNO2
l-2-bromophenylalanine, 2-bromo-l-phenylalanine, s-2-amino-3-2-bromophenyl propanoic acid, 2-bromophenylalanine, 2s-2-amino-3-2-bromophenyl propanoic acid, h-phe 2-br-oh, o-bromo-l-phenylalanine, l-2-br-phe-oh, s-2-bromophenylalanine, l-2-bromophe
C1=CC=C(C(=C1)CC(C(=O)O)N)Br
244.088
244.09
95%
250mg
42538-40-9
MFCD03092958
JFVLNTLXEZDFHW-QMMMGPOBSA-N
(2S)-2-amino-3-(2-bromophenyl)propanoic acid
193338
Powder
White
Videos
Documents