Learn More
2-Bromo-N-[4-chloro-2-(trifluoroacetyl)phenyl]acetamide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-N-[4-chloro-2-(trifluoroacetyl)phenyl]acetamide |
| CAS | 253668-48-3 |
| Molecular Formula | C10H6BrClF3NO2 |
| Quantity | 100 mg |
| Synonym | 2-Bromo-N-[4-chloro-2-(2,2,2-trifluoroacetyl)phenyl]acetamide |
| InChI Formula | InChI=1S/C10H6BrClF3NO2/c11-4-8(17)16-7-2-1-5(12)3-6(7)9(18)10(13,14)15/h1-3H,4H2,(H,16,17) |
| SMILES | ClC1=CC=C(NC(CBr)=O)C(C(C(F)(F)F)=O)=C1 |
| IUPAC Name | 2-bromo-N-(4-chloro-2-(2,2,2-trifluoroacetyl)phenyl)acetamide |
| Molecular Weight (g/mol) | 344.51 |
| Formula Weight | 342.92 |