missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromoacetophenone, 98+%, Thermo Scientific Chemicals

Catalog Number p-4509496
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2142-69-0
C8H7BrO
199.05
MFCD00000067
PIMNFNXBTGPCIL-UHFFFAOYSA-N
2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone
75060
1-(2-bromophenyl)ethanone
CC(=O)C1=CC=CC=C1Br
This item is not returnable. View return policy
2142-69-0
C8H7BrO
199.05
MFCD00000067
PIMNFNXBTGPCIL-UHFFFAOYSA-N
2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone
75060
1-(2-bromophenyl)ethanone
CC(=O)C1=CC=CC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Orange to Yellow
Density 1.4760g/mL
Boiling Point 116.0°C to 117.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4COCH3
Refractive Index 1.5650 to 1.5690
Quantity 5 g
Specific Gravity 1.476
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: freely soluble in alcohol,benzene,chlorofom,eth
Formula Weight 199.05
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2'-Bromoacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2'-Bromoacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'