Learn More
2-Bromoethyl Methanethiosulfonate, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromoethyl Methanethiosulfonate |
| CAS | 26139-15-1 |
| Molecular Formula | C3 H7 Br O2 S2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C3H7BrO2S2/c1-8(5,6)7-3-2-4/h2-3H2,1H3 |
| SMILES | CS(=O)(=O)SCCBr |
| IUPAC Name | 1-bromo-2-methylsulfonylsulfanylethane |
| Molecular Weight (g/mol) | 219.12 |
| Formula Weight | 217.9071 |