Learn More
2-Bromoethyl Methanethiosulfonate, TRC
Product Code. 30344178
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30344178 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30344178 Supplier Toronto Research Chemicals Supplier No. TRCB684250100MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Bromoethyl Methanethiosulfonate
CAS 26139-15-1
Molecular Formula C3 H7 Br O2 S2
Quantity 100 mg
InChI Formula InChI=1S/C3H7BrO2S2/c1-8(5,6)7-3-2-4/h2-3H2,1H3
SMILES CS(=O)(=O)SCCBr
IUPAC Name 1-bromo-2-methylsulfonylsulfanylethane
Molecular Weight (g/mol) 219.12
Formula Weight 217.9071