Learn More
2-Butanone 2,4-Dinitrophenylhydrazone-3,5,6-d3, CDN
Product Code. 30311975
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311975 0.01 g 0.01g
30312130 0.005 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30311975 Supplier CDN Supplier No. CDND76020.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Butanone 2,4-Dinitrophenylhydrazone-d3
CAS 259824-58-3
Molecular Formula C10 D3 H9 N4 O4
Quantity 0.01 g
Synonym 2-Butanone D3 (phenyl D3) 2,4-Dinitrophenylhydrazone, 2-Butanone, (4,6-dinitrophenyl-2,3,5-d3)hydrazone (9 CI)
InChI Formula InChI=1 S/C10H12N4O4/c1-3-7(2)11-12-9-5-4-8(13(15)16)6-10(9)14(17)18/h4-6,12 H,3H2,1-2H3/b11-7+/i4D,5 D,6 D
SMILES [2 H]c1c([2 H])c(c([2 H])c(c1N\N=C(/C)\CC)[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name N-[(E)-butan-2-ylideneamino]-2,3,5-trideuterio-4,6-dinitroaniline
Molecular Weight (g/mol) 255.245
Formula Weight 255.105 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity