Learn More
2-Butanone 2,4-Dinitrophenylhydrazone-d3, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Butanone 2,4-Dinitrophenylhydrazone-d3 |
| CAS | 259824-58-3 |
| Molecular Formula | C10 D3 H9 N4 O4 |
| Quantity | 50 mg |
| Synonym | 2-Butanone D3 (phenyl D3) 2,4-Dinitrophenylhydrazone, 2-Butanone, (4,6-dinitrophenyl-2,3,5-d3)hydrazone (9CI) |
| InChI Formula | InChI=1S/C10H12N4O4/c1-3-7(2)11-12-9-5-4-8(13(15)16)6-10(9)14(17)18/h4-6,12H,3H2,1-2H3/b11-7+/i4D,5D,6D |
| SMILES | [2H]c1c([2H])c(c([2H])c(c1N\N=C(/C)\CC)[N+](=O)[O-])[N+](=O)[O-] |
| IUPAC Name | N-[(E)-butan-2-ylideneamino]-2,3,5-trideuterio-4,6-dinitroaniline |
| Molecular Weight (g/mol) | 255.245 |
| Formula Weight | 255.105 |