missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Butylbenzofuran-3-yl)(4-methoxyphenyl)methanone, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
(2-Butylbenzofuran-3-yl)(4-methoxyphenyl)methanone
Specifications
Specifications
| Chemical Name or Material | De(diethylaminomethyldiiodo) Amiodarone |
| CAS | 83790-87-8 |
| Analyte or Component Names | De(diethylaminomethyldiiodo) Amiodarone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H20 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Methanone, (2-butyl-3-benzofuranyl)(4-methoxyphenyl)-, Ketone, 2-butyl-3-benzofuranyl p-methoxyphenyl (7CI), (2-Butyl-3-benzofuranyl)(4-methoxyphenyl)methanone, (2-Butylbenzofuran-3-yl)(4-methoxyphenyl)methanone, 2-Butyl-3-benzofuranyl p-Methoxyphenyl Ketone, (2-Butylbenzofuran-3-yl)(4-methoxyphenyl)methanone, (2-Butylbenzofuran-3-yl)(4-methoxyphenyl)methanone |
| InChI Formula | InChI=1S/C20H20O3/c1-3-4-8-18-19(16-7-5-6-9-17(16)23-18)20(21)14-10-12-15(22-2)13-11-14/h5-7,9-13H,3-4,8H2,1-2H3 |
| SMILES | CCCCc1oc2ccccc2c1C(=O)c3ccc(OC)cc3 |
| IUPAC Name | (2-butyl-1-benzofuran-3-yl)-(4-methoxyphenyl)methanone |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction