missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Carbaminidoylsulfanyl)ethanesulfonic Acid, Mikromol™
Description
2-(Carbaminidoylsulfanyl)ethanesulfonic Acid, Mikromol
Specifications
Specifications
| Chemical Name or Material | 2-(Carbaminidoylsulfanyl)ethanesulfonic Acid |
| CAS | 25985-57-3 |
| Analyte or Component Names | 2-(Carbaminidoylsulfanyl)ethanesulfonic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C3 H8 N2 O3 S2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Mesna Imp. A (EP), 2-[(Aminoiminomethyl)thio]ethanesulfonic Acid, Ethanesulfonic acid, 2-(amidinothio)- (6CI,7CI,8CI), Pseudourea, 2-(2-sulfoethyl)- (8CI), 2-S-Thiuronium ethanesulfonate, Carbamimidothioic acid, 2-sulfoethyl ester, 2-(Carbaminidoylsulfanyl)ethanesulfonic Acid, Mesna Impurity A |
| Solution Type | Neat |
| SMILES | NC(=N)SCCS(=O)(=O)O |
| Molecular Weight (g/mol) | 184.23 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?