Learn More
(2-Carboxyethyl)triphenylphosphonium chloride, 98%
Product Code. 11461637
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461637 100 g 100g
11441637 5 g 5g
11451637 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11461637 Supplier Thermo Scientific Alfa Aesar Supplier No. B25609.22
Request Bulk or Custom Format
CAS: 36626-29-6 | C21H20ClO2P | 370.81 g/mol
2-Carboxyethyl)-triphenylphosphonium chloride is a phosphonium ylide reagent for Wittig olefinations with introduction of a carboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Carboxyethyl)-triphenylphosphonium chloride is a phosphonium ylide reagent for Wittig olefinations with introduction of a carboxylic acid.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 36626-29-6
Molecular Formula C21H20ClO2P
Molecular Weight (g/mol) 370.81
MDL Number MFCD00185663
InChI Key GALLWJZTZYJVSL-UHFFFAOYSA-N
Synonym 2-carboxyethyl triphenylphosphonium chloride, 2-carboxyethyl tripheylphosphonium chloride, 2-carboxyethyl-triphenylphosphonium chloride, phosphonium, 2-carboxyethyl triphenyl-, chloride, 2-carboxyethyl triphenylphosphanium chloride, acmc-1agne, 2-carboxyethyl triphenyl phosphonium chloride, beta-carboxyethyltriphenylphosphonium chloride, 2-carboxyethyl triphenyl-phosphonium chloride
PubChem CID 10270534
SMILES [Cl-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point 197°C to 201°C
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in dimethyl sulfoxide and methanol.
IUPAC Name 2-carboxyethyl(triphenyl)phosphanium;chloride
Formula Weight 370.81
Percent Purity 98%
Chemical Name or Material (2-Carboxyethyl)triphenylphosphonium chloride