Learn More
2-Chloro-1,5-hexadiene (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-1,5-hexadiene |
| CAS | 101933-88-4 |
| Molecular Formula | C6H9Cl |
| Quantity | 25 mg |
| Synonym | 2-Chlorohexa-1,5-Diene |
| InChI Formula | InChI=1S/C6H9Cl/c1-3-4-5-6(2)7/h3H,1-2,4-5H2 |
| SMILES | ClC(CCC=C)=C |
| IUPAC Name | 2-Chloro-1,5-hexadiene (ACI) |
| Molecular Weight (g/mol) | 116.59 |
| Formula Weight | 116.04 |