missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,5-difluorophenol, 98%, Thermo Scientific™

Catalog Number p-4228498
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
206986-81-4
C6H3ClF2O
164.54
ZQWULOHSTAQTKJ-UHFFFAOYSA-N
2733297
2-chloro-3,5-difluorophenol
C1=C(C=C(C(=C1F)Cl)O)F
This item is not returnable. View return policy
206986-81-4
C6H3ClF2O
164.54
ZQWULOHSTAQTKJ-UHFFFAOYSA-N
2733297
2-chloro-3,5-difluorophenol
C1=C(C=C(C(=C1F)Cl)O)F
CAS Min % 97.5
CAS Max % 100.0
Melting Point 45.0°C to 46.0°C
Color White
Boiling Point 76.0°C (37.0 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Synonym phenol, 2-chloro-3,5-difluoro, acmc-1cs4q, 2-chloro-3,5-difluoro-phenol, phenol,2-chloro-3,5-difluoro, 4,6-difluoro-2-hydroxychlorobenzene, 2-chloro-3,5-difluorophenol
Formula Weight 164.54
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-3, 5-difluorophenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-3,5-difluorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'