missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,5-dimethylphenol, Mikromol™

Product Code. 30052204
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052204 25 mg 25mg
30051606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052204 Supplier Mikromol Supplier No. MM1997.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Chloro-3,5-dimethylphenol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-3,5-dimethylphenol
CAS 5538-41-0
Analyte or Component Names 2-Chloro-3,5-dimethylphenol
Shipping Condition Room Temperature
Molecular Formula C8 H9 Cl O
Certifications/Compliance ISO 17025
Synonym Phenol, 2-chloro-3,5-dimethyl-, 3,5-Xylenol, 2-chloro- (7CI,8CI), 2-Chloro-3,5-dimethylphenol, 2-Chloro-3,5-xylenol, Chloroxylenol Related Compound A
InChI Formula InChI=1S/C8H9ClO/c1-5-3-6(2)8(9)7(10)4-5/h3-4,10H,1-2H3
SMILES Cc1cc(C)c(Cl)c(O)c1
IUPAC Name 2-chloro-3,5-dimethylphenol
Molecular Weight (g/mol) 156.61
Quantity 25 mg
Formula Weight 156.0342 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.