Learn More
2-Chloro-3,5-dinitropyridine, 98+%, Thermo Scientific™
Product Code. 10432021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10432021 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10432021 Supplier Thermo Scientific Acros Supplier No. 208190010
Request Bulk or Custom Format
Specifications
CAS 2578-45-2
CAS Min % 98.5
CAS Max % 100.0
Melting Point 63.0°C to 66.0°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C5H2ClN3O4
Linear Formula (O2H)(C5H2N)Cl
MDL Number MFCD00006233
Quantity 1 g
Beilstein 20,III,2536
Synonym pyridine, 2-chloro-3,5-dinitro, 2-chloro-3,5-dinitro-pyridine, pubchem6103, acmc-209gmh, 2-chloro3,5-dinitropyridine, 2-chloro-3,5-dinitropyridine
InChI Key QLHVJBXAQWPEDI-UHFFFAOYSA-N
SMILES C1=C(C=NC(=C1[N+](=O)[O-])Cl)[N+](=O)[O-]
IUPAC Name 2-chloro-3,5-dinitropyridine
Molecular Weight (g/mol) 203.54
PubChem CID 75738
Formula Weight 203.54
Percent Purity 98+%
Chemical Name or Material 2-Chloro-3,5-dinitropyridine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-3,5-dinitropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.