Learn More
2-Chloro-3,5-dinitropyridine, 98+%
Product Code. 13478458 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13478458 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13478458 Supplier Thermo Scientific Acros Supplier No. 208190250
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 2578-45-2 | C5H2ClN3O4 | 203.54 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 2578-45-2
CAS Min % 98.5
CAS Max % 100.0
Melting Point 63.0°C to 66.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C5H2ClN3O4
Linear Formula (O2H)(C5H2N)Cl
MDL Number MFCD00006233
Quantity 25 g
Beilstein 20,III,2536
Synonym pyridine, 2-chloro-3,5-dinitro, 2-chloro-3,5-dinitro-pyridine, pubchem6103, acmc-209gmh, 2-chloro3,5-dinitropyridine, 2-chloro-3,5-dinitropyridine
InChI Key QLHVJBXAQWPEDI-UHFFFAOYSA-N
SMILES C1=C(C=NC(=C1[N+](=O)[O-])Cl)[N+](=O)[O-]
IUPAC Name 2-chloro-3,5-dinitropyridine
Molecular Weight (g/mol) 203.54
PubChem CID 75738
Formula Weight 203.54
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3,5-dinitropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-3,5-dinitropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.