Learn More
2-Chloro-3-bromo-5-methylpyridine, ≥97%, Thermo Scientific™
Product Code. 10524763
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10524763 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10524763 Supplier Thermo Scientific Maybridge Supplier No. MO07184DA
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Specifications
CAS 17282-03-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H5BrClN
MDL Number MFCD01830664
Quantity 1 g
Synonym 2-chloro-3-bromo-5-methylpyridine, 3-bromo-2-chloro-5-picoline, 5-bromo-6-chloro-3-picoline, pyridine, 3-bromo-2-chloro-5-methyl, 3-bromo-2-chloro-5-methyl-pyridine, 2-chloro-5-methyl-3-bromopyridine, pubchem1167, acmc-1cbw1, ksc490s4b, 2-chloro-3-bromo-5-picoline
InChI Key ISNRJFOYJNIKTN-UHFFFAOYSA-N
SMILES CC1=CC(=C(N=C1)Cl)Br
IUPAC Name 3-bromo-2-chloro-5-methylpyridine
Molecular Weight (g/mol) 206.467
PubChem CID 285437
Formula Weight 206.47
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-bromo-5-methylpyridine