missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluoropyridine, 97%, Thermo Scientific™

Catalog Number 11966421
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
17282-04-1
C5H3ClFN
131.54
SVAZIMBLBHOVIR-UHFFFAOYSA-N
3-fluoro-2-chloropyridine, 2-chloro-3-fluoro pyridine, pyridine, 2-chloro-3-fluoro, 2-chloro-3-fluoro-pyridine, pubchem2532, acmc-1cc0d, ksc174s3h, 2-chloro-3-fluoropyridine, 2-chloro-3-fluoropyridine 1g
2782791
2-chloro-3-fluoropyridine
C1=CC(=C(N=C1)Cl)F
This item is not returnable. View return policy
17282-04-1
C5H3ClFN
131.54
SVAZIMBLBHOVIR-UHFFFAOYSA-N
3-fluoro-2-chloropyridine, 2-chloro-3-fluoro pyridine, pyridine, 2-chloro-3-fluoro, 2-chloro-3-fluoro-pyridine, pubchem2532, acmc-1cc0d, ksc174s3h, 2-chloro-3-fluoropyridine, 2-chloro-3-fluoropyridine 1g
2782791
2-chloro-3-fluoropyridine
C1=CC(=C(N=C1)Cl)F
Density 1.3230g/mL
Boiling Point 80°C (80.0 mmHg)
Flash Point 63°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.323
Formula Weight 131.54
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-fluoropyridine
compliance-icons
Product Identifier
  • 2-Chloro-3-fluoropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'