Learn More
2-Chloro-4-fluoro-3-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Product Code. 15458377
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458377 1 g 1g
15468377 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15458377 Supplier Thermo Scientific Alfa Aesar Supplier No. H51879.03
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 173315-54-3 | Molecular Formula: C8H6ClFO2 | Molecular Weight: 188.58 g/mol
It is used in the synthesis and Antibacterial Evaluation of 6-Amino-8-methylquinolones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the synthesis and Antibacterial Evaluation of 6-Amino-8-methylquinolones.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 173315-54-3
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.58
MDL Number MFCD11846444
InChI Key DBMUFFHLBXKTDM-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-4-fluoro-3-methyl, 2-chloro-4-fluoro-3-methyl-benzoic acid, 2-chloro-3-methyl-4-fluoro-benzoic acid, 2-chloro-4-fluoro-3-methyl benzoic acid
PubChem CID 10845315
IUPAC Name 2-chloro-4-fluoro-3-methylbenzoic acid
SMILES CC1=C(F)C=CC(C(O)=O)=C1Cl
Specifications
Melting Point 186°C to 189°C
Quantity 1 g
Beilstein 7423351
Formula Weight 188.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluoro-3-methylbenzoic acid
RUO – Research Use Only
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.