missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-(2,3-dimethylphenyl)benzamide, 97%

Product Code. 15452668
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15452668

Brand: Thermo Scientific Alfa Aesar H56225.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 196617-88-6 | C15H14ClNO | 259.733 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 196617-88-6
Molecular Formula C15H14ClNO
Molecular Weight (g/mol) 259.733
MDL Number MFCD00018254
InChI Key OUAGJVUPQKYHDU-UHFFFAOYSA-N
Synonym 2-chloro-n-2,3-dimethylphenyl benzamide, cambridge id 5328590, 2-chloro-2',3'-benzoxylidide, 2-chloro-n-2,3-dimethyl-phenyl-benzamide, n-2,3-dimethylphenyl 2-chlorophenyl carboxamide
PubChem CID 295079
IUPAC Name 2-chloro-N-(2,3-dimethylphenyl)benzamide
SMILES CC1=C(C(=CC=C1)NC(=O)C2=CC=CC=C2Cl)C

Specifications

Quantity 1 g
Formula Weight 259.74
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-(2,3-dimethylphenyl)benzamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.