missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-[2-(diethylamino)ethyl]-4-quinolinecarboxamide, TRC

Product Code. 30008853
Change view
Click to view available options
Quantity:
500 mg
10 g
Unit Size:
10g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008853 500 mg 500mg
30008394 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008853 Supplier Toronto Research Chemicals Supplier No. TRCC365260500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87864-14-0
Molecular Formula C16 H20 Cl N3 O
Molecular Weight (g/mol) 305.8
Synonym 4-Quinolinecarboxamide, 2-chloro-N-[2-(diethylamino)ethyl]-, Cinchoninamide, 2-chloro-N-(2-diethylaminoethyl)- (6CI)
IUPAC Name 2-chloro-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide
SMILES CCN(CC)CCNC(=O)c1cc(Cl)nc2ccccc12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H20ClN3O/c1-3-20(4-2)10-9-18-16(21)13-11-15(17)19-14-8-6-5-7-12(13)14/h5-8,11H,3-4,9-10H2,1-2H3,(H,18,21)
Formula Weight 305.1295
Chemical Name or Material 2-Chloro-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.