Learn More
2-Chloro-N-(phenylmethyl)-1H-purin-6-amine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-N-(phenylmethyl)-1H-purin-6-amine |
| CAS | 39639-47-9 |
| Molecular Formula | C12H10ClN5 |
| Quantity | 1 g |
| Synonym | 6-Benzylamino-2-chloropurine, 2-Chloro-N-(phenylmethyl)-1H-purin-6-amine |
| InChI Formula | InChI=1S/C12H10ClN5/c13-12-17-10(9-11(18-12)16-7-15-9)14-6-8-4-2-1-3-5-8/h1-5,7H,6H2,(H2,14,15,16,17,18) |
| SMILES | ClC1=NC(NC=N2)=C2C(NCC3=CC=CC=C3)=N1 |
| IUPAC Name | N-benzyl-2-chloro-9H-purin-6-amine |
| Molecular Weight (g/mol) | 259.69 |
| Formula Weight | 259.06 |