missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethylphosphonic acid, 90%, Tech.

Product Code. 10227750 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10227750

Brand: Thermo Scientific Acros 301630050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 16672-87-0 | C2H6ClO3P | 144.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 16672-87-0
Molecular Formula C2H6ClO3P
Molecular Weight (g/mol) 144.49
MDL Number MFCD00002142
InChI Key UDPGUMQDCGORJQ-UHFFFAOYSA-N
Synonym ethephon, 2-chloroethyl phosphonic acid, ethrel, chlorethephon, bromeflor, camposan, 2-cepa, phosphonic acid, 2-chloroethyl, 2-chloroethanephosphonic acid, etheverse
PubChem CID 27982
ChEBI CHEBI:52741
IUPAC Name 2-chloroethylphosphonic acid
SMILES OP(O)(=O)CCCl

Spécification

CAS Min % 88.0
CAS Max % 100.0
Melting Point 74.0°C to 75.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 3787
Solubility Information Solubility in water: freely soluble. Other solubilities: slightly soluble in benzene,toluene,propylene glycol,practically insoluble in petroleum ether,practically insoluble in petroleum ether,freely soluble in ethanol,methanol,acetone,ethy
Formula Weight 144.51
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material 2-Chloroethylphosphonic acid, tech., 90%
Afficher plus Afficher moins

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis