missing translation for 'onlineSavingsMsg'
Learn More

2-chloropyrimidine-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4228536
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
374068-01-6
C5H3ClN2O2
158.54
MFCD06739036
DUCXUPKLVVSJKA-UHFFFAOYSA-N
2-chloro-pyrimidine-5-carboxylic acid, 2-chloro-5-carboxypyrimidine, 5-carboxy-2-chloropyrimidine, 5-pyrimidinecarboxylicacid, 2-chloro, 2-chloro-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylic acid, 2-chloro, 2-chloropyrimidine-5-carboxylicacid, acmc-1aig3, 2-chloropyrimidine 5-carboxylic acid, 2-chloro-5-pyrimidine carboxylic acid
11094906
2-chloropyrimidine-5-carboxylic acid
OC(=O)C1=CN=C(Cl)N=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
374068-01-6
C5H3ClN2O2
158.54
MFCD06739036
DUCXUPKLVVSJKA-UHFFFAOYSA-N
2-chloro-pyrimidine-5-carboxylic acid, 2-chloro-5-carboxypyrimidine, 5-carboxy-2-chloropyrimidine, 5-pyrimidinecarboxylicacid, 2-chloro, 2-chloro-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylic acid, 2-chloro, 2-chloropyrimidine-5-carboxylicacid, acmc-1aig3, 2-chloropyrimidine 5-carboxylic acid, 2-chloro-5-pyrimidine carboxylic acid
11094906
2-chloropyrimidine-5-carboxylic acid
OC(=O)C1=CN=C(Cl)N=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 125.0°C to 127.5°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 158.54
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Chloropyrimidine-5-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloropyrimidine-5-carboxylic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'