Learn More
Thermo Scientific Alfa Aesar 50ML 2-Cyanoethylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal^t bottles 5
Brand:  Thermo Scientific Alfa Aesar H58695.AD
50ML 2-Cyanoethylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal^t bottles 5
Additional Details : CAS : 312624-26-3 Weight : 0.00050kg Transport : UN number : 3399 Chem class : 4.3 Pack group : II
Product Code. 15495395