Learn More
2’-Deoxy-2’-fluoro Cytidine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 2'-Deoxy-2'-fluoro Cytidine |
| CAS | 10212-20-1 |
| Molecular Formula | C9 H12 F N3 O4 |
| Quantity | 1 g |
| Synonym | Cytidine, 2'-deoxy-2'-fluoro-, 2'-Deoxy-2'-fluorocytidine, 1562: PN: US6617438 TABLE: 1 claimed sequence, 2'-Fluoro-2'-deoxy-β-D-ribofuranosylcytosine, 2'-Fluoro-2'-deoxycytidine, NSC 529432 |
| InChI Formula | InChI=1S/C9H12FN3O4/c10-6-7(15)4(3-14)17-8(6)13-2-1-5(11)12-9(13)16/h1-2,4,6-8,14-15H,3H2,(H2,11,12,16)/t4-,6-,7-,8-/m1/s1 |
| SMILES | NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2F |
| IUPAC Name | 4-amino-1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
| Molecular Weight (g/mol) | 245.21 |
| Formula Weight | 245.0812 |