Learn More
2-Ethyl-6-methoxynaphthalene (Ethylnerolin), Mikromol™
Product Code. 17992766
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17992766 100 mg 100mg
17960586 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17992766 Supplier Mikromol Supplier No. MM0152.18
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Ethyl-6-methoxynaphthalene (Ethylnerolin)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Ethyl-6-methoxynaphthalene (Ethylnerolin)
CAS 21388-17-0
Analyte or Component Names 2-Ethyl-6-methoxynaphthalene (Ethylnerolin)
Shipping Condition Room Temperature
Molecular Formula C13 H14 O
Certifications/Compliance ISO 17025
Synonym 2-Ethyl-6-methoxynaphthalene, Naproxen Imp. J (EP), Ethylnerolin, Naproxen Impurity J
InChI Formula InChI=1S/C13H14O/c1-3-10-4-5-12-9-13(14-2)7-6-11(12)8-10/h4-9H,3H2,1-2H3
SMILES CCc1ccc2cc(OC)ccc2c1
IUPAC Name 2-ethyl-6-methoxynaphthalene
Molecular Weight (g/mol) 186.25
Quantity 100 mg
Formula Weight 186.1045 g/mol