Learn More
2-Ethylhexanoic Acid Lithium Salt, TRC
Product Code. 30385038
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385038 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30385038 Supplier Toronto Research Chemicals Supplier No. TRCE1559052.5MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Chemical Name or Material 2-Ethylhexanoic Acid Lithium Salt
CAS 15590-62-2
Molecular Formula C8H15LiO2
Quantity 2.5 mg
Synonym Lithium 2-ethylhexanoate, NUSA Li
InChI Formula InChI=1S/C8H16O2.Li.H/c1-3-5-6-7(4-2)8(9)10;;/h7H,3-6H2,1-2H3,(H,9,10);;/p-1
SMILES [Li].CCCCC(CC)C(=O)[O-]
IUPAC Name 2-ethylhexanoate;lithium hydride
Molecular Weight (g/mol) 150.14
Formula Weight 150.1232
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.