missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-heptyl sulfate, 99% (dry wt.), water <2%, Thermo Scientific™

Product Code. 11409617
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11409617

Brand: Thermo Scientific Alfa Aesar B22104.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18981-98-1
Molecular Formula C7H16NaO4S
Molecular Weight (g/mol) 219.251
MDL Number MFCD00051524
InChI Key SLGMFWJQQBPOMZ-UHFFFAOYSA-N
Synonym heptyl sodium sulfate
PubChem CID 131875435
IUPAC Name heptyl hydrogen sulfate;sodium
SMILES CCCCCCCOS(=O)(=O)O.[Na]

Specifications

Melting Point 199°C to 202°C (decomposition)
Linear Formula CH3(CH2)6OSO3Na
Quantity 5 g
Beilstein 6018116
Formula Weight 218.24
Percent Purity 99%
Assay (dry wt.), water <2%
Chemical Name or Material Sodium n-heptyl sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.