missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-1,4-bis(trifluoromethyl)benzene, 95%, Thermo Scientific™

Product Code. 15426057
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15426057 5 g 5g
15436057 250 mg 250mg
15416057 1 g 1g
3 options
This item is not returnable. View return policy

Product Code. 15426057

Brand: Thermo Scientific Alfa Aesar H33241.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 887268-08-8
Molecular Formula C8H3F7
Molecular Weight (g/mol) 232.10
MDL Number MFCD03412236
InChI Key KAGRJEBZBWYSJK-UHFFFAOYSA-N
Synonym 2,5-bis trifluoromethyl fluorobenzene, 2-fluoro-1,4-bis-trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-fluorobenzene, 2-fluoro-1,4-bis trifluoromethyl benzene
PubChem CID 44717336
IUPAC Name 2-fluoro-1,4-bis(trifluoromethyl)benzene
SMILES FC1=C(C=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Boiling Point 50°C to 54°C (60 mmHg)
Quantity 5 g
Formula Weight 232.09
Percent Purity 95%
Chemical Name or Material 2-Fluoro-1,4-bis(trifluoromethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.