missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxyphenylboronic acid, 97%, ACROS Organics™

Brand:  Acros Organics 449840010

Additional Details : CAS : 162101-31-7 Weight : 0.00100kg


Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 15224784

  • € 38.38 / 1g


Estimated Shipment
01-06-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

162101-31-7
169.95
2-fluoro-4-methoxybenzeneboronic acid, 2-fluoro-4-methoxyphenyl boronic acid, 4-borono-3-fluoroanisole, boronic acid, 2-fluoro-4-methoxyphenyl, boronic acid, b-2-fluoro-4-methoxyphenyl, 2-fluoro-4-methoxyphenylboronicacid, pubchem15367, acmc-209dnv, ksc498a9p, 2-fluoro-4methoxyphenylboronic acid
(2-fluoro-4-methoxyphenyl)boronic acid
C7H8BFO3
ULUIXJDBPYBAHS-UHFFFAOYSA-N
15789384
B(C1=C(C=C(C=C1)OC)F)(O)O

Specifications

2-Fluoro-4-methoxyphenylboronic acid
1g
96% min. (HPLC)
2-fluoro-4-methoxybenzeneboronic acid, 2-fluoro-4-methoxyphenyl boronic acid, 4-borono-3-fluoroanisole, boronic acid, 2-fluoro-4-methoxyphenyl, boronic acid, b-2-fluoro-4-methoxyphenyl, 2-fluoro-4-methoxyphenylboronicacid, pubchem15367, acmc-209dnv, ksc498a9p, 2-fluoro-4methoxyphenylboronic acid
B(C1=C(C=C(C=C1)OC)F)(O)O
169.95
169.95
Glass bottle
162101-31-7
C7H8BFO3
ULUIXJDBPYBAHS-UHFFFAOYSA-N
(2-fluoro-4-methoxyphenyl)boronic acid
15789384
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Fluoro-4-methoxyphenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents