missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-N-methylbenzylamine, 95%, Thermo Scientific Chemicals

Catalog Number p-7052628
29.93 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
399-30-4
C8H10FN
139.173
MFCD04629633
AHIHZCXUWGORQO-UHFFFAOYSA-N
n-methyl-2-fluorobenzylamine, 2-fluorobenzyl methylamine, 2-fluoro-n-methylbenzylamine, 2-fluorophenyl methyl methyl amine, 1-2-fluorophenyl-n-methylmethanamine, n-2-fluorobenzyl-n-methylamine, n-methyl-o-fluorobenzylamine, benzenemethanamine, 2-fluoro-n-methyl, 1-2-fluorophenyl-n-methyl-methanamine, acmc-209zio
2060842
1-(2-fluorophenyl)-N-methylmethanamine
CNCC1=CC=CC=C1F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
399-30-4
C8H10FN
139.173
MFCD04629633
AHIHZCXUWGORQO-UHFFFAOYSA-N
n-methyl-2-fluorobenzylamine, 2-fluorobenzyl methylamine, 2-fluoro-n-methylbenzylamine, 2-fluorophenyl methyl methyl amine, 1-2-fluorophenyl-n-methylmethanamine, n-2-fluorobenzyl-n-methylamine, n-methyl-o-fluorobenzylamine, benzenemethanamine, 2-fluoro-n-methyl, 1-2-fluorophenyl-n-methyl-methanamine, acmc-209zio
2060842
1-(2-fluorophenyl)-N-methylmethanamine
CNCC1=CC=CC=C1F

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and air. Store at 4-2°C.
TRUSTED_SUSTAINABILITY
Color Colorless to Tan
Density 1.047 g/mL
Boiling Point 73°C
Flash Point 68°C (154°F)
Refractive Index 1.047
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 139.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-N-methylbenzylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'