Learn More
2-fluoropropanedioic acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-fluoropropanedioic acid |
| CAS | 473-87-0 |
| Molecular Formula | C3H3O4F |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C3H3FO4/c4-1(2(5)6)3(7)8/h1H,(H,5,6)(H,7,8) |
| SMILES | C(C(=O)O)(C(=O)O)F |
| IUPAC Name | 2-fluoropropanedioic acid |
| Molecular Weight (g/mol) | 122.05 |
| Formula Weight | 122.0015 |