missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazinopyrazine, 98%, Thermo Scientific Chemicals

Catalog Number 15441129
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
54608-52-5
C4H6N4
110.12
MFCD04114555
IVRLZJDPKUSDCF-UHFFFAOYSA-N
2-hydrazinylpyrazine, 2-hydrazinopyrazine, pyrazin-2-yl-hydrazine, 1-pyrazin-2-yl hydrazine, hydrazinopyrazine, 2-pyrazin-2-yl hydrazine, 2 1h-pyrazinone, hydrazone, hydrazino pyrazine, pyrazinyl hydrazine, 3-hydrazinopyrazine
1487823
pyrazin-2-ylhydrazine
C1=CN=C(C=N1)NN
This item is not returnable. View return policy
54608-52-5
C4H6N4
110.12
MFCD04114555
IVRLZJDPKUSDCF-UHFFFAOYSA-N
2-hydrazinylpyrazine, 2-hydrazinopyrazine, pyrazin-2-yl-hydrazine, 1-pyrazin-2-yl hydrazine, hydrazinopyrazine, 2-pyrazin-2-yl hydrazine, 2 1h-pyrazinone, hydrazone, hydrazino pyrazine, pyrazinyl hydrazine, 3-hydrazinopyrazine
1487823
pyrazin-2-ylhydrazine
C1=CN=C(C=N1)NN

Triazolo[4,3-a]pyrazine 7-oxide, 3-β-D-ribofuranosyl-1,2,4-triazolo[4,3-a]pyrazine, monohydrazones of pyridine, pyrazine, phenylglyoxal were synthesised from 2-Hydrazinopyrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triazolo[4,3-a]pyrazine 7-oxide, 3-β-D-ribofuranosyl-1,2,4-triazolo[4,3-a]pyrazine, monohydrazones of pyridine, pyrazine, phenylglyoxal were synthesised from 2-Hydrazinopyrazine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 109°C to 113°C
Color Yellow
Quantity 250 mg
Beilstein 606630
Solubility Information Slightly soluble in water.
Formula Weight 110.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Hydrazinopyrazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'