Learn More
2-Hydroxy-1-methyl-6-phenylimidazo(4,5-b)pyridine-d3, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy-1-(trideuteromethyl)-6-phenylimidazo(4,5-b)pyridin |
| CAS | 1020719-48-5 |
| Molecular Formula | C13 2H3 H8 N3 O |
| Quantity | 1 mg |
| Synonym | 2H-Imidazo[4,5-b]pyridin-2-one, 1,3-dihydro-1-(methyl-d3)-6-phenyl-, 1,3-Dihydro-1-trideuteriomethyl-6-phenyl-2H-imidazo[4,5-b]pyridin-2-one, 2-Hydroxy-1-methyl-6-phenylimidazo[4,5-b]pyridine-D3 (N-methyl-D3), 1-(Methyl-d3)-6-phenyl-1H-imidazo[4,5-b]pyridin-2-ol |
| InChI Formula | InChI=1S/C13H11N3O/c1-16-11-7-10(9-5-3-2-4-6-9)8-14-12(11)15-13(16)17/h2-8H,1H3,(H,14,15,17)/i1D3 |
| SMILES | [2H]C([2H])([2H])n1c(O)nc2ncc(cc12)c3ccccc3 |
| IUPAC Name | 6-phenyl-1-(trideuteriomethyl)imidazo[4,5-b]pyridin-2-ol |
| Molecular Weight (g/mol) | 228.26 |
| Formula Weight | 228.109 |