Learn More
2-Hydroxy-4-methylpyridine, 98+%
Product Code. 11332927
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332927 5 g 5g
11322927 1 g 1g
11342927 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11332927 Supplier Thermo Scientific Alfa Aesar Supplier No. L16188.06
Request Bulk or Custom Format
CAS: 13466-41-6 | C6H7NO | 109.13 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13466-41-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00134298,MFCD00030766
InChI Key YBDRFJXGJQULGH-UHFFFAOYSA-N
Synonym 2-hydroxy-4-methylpyridine, 4-methylpyridin-2-ol, 4-methyl-2 1h-pyridinone, 2 1h-pyridinone, 4-methyl, 2-hydroxy-4-picoline, 4-methyl-2-pyridinol, 4-methyl-1,2-dihydropyridin-2-one, 4-methyl-2-pyridone, 4-methylpyridin-2 1h-one, 2-hydroxy-4-methyl pyridine
PubChem CID 280741
SMILES CC1=CC(=O)NC=C1
Specifications
Melting Point 128°C to 131°C
Boiling Point 186°C to 187°C (12 mmHg)
Odor Odorless
Quantity 5 g
Beilstein 107082
IUPAC Name 4-methyl-1,2-dihydropyridin-2-one
Formula Weight 109.13
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxy-4-methylpyridine