missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-(octyloxy)benzophenone, TRC

Product Code. 30005986
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005986 5 g 5g
30135116 100 g 100g
30005337 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005986 Supplier Toronto Research Chemicals Supplier No. TRCH9510985G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1843-05-6
Molecular Formula C21 H26 O3
Molecular Weight (g/mol) 326.43
Synonym Benzophenone, 2-hydroxy-4-(octyloxy)- (6CI,8CI), [2-Hydroxy-4-(octyloxy)phenyl]phenylmethanone, 2-Benzoyl-5-octyloxyphenol, 2-Hydroxy-4-(n-octyloxy)benzophenone, 2-Hydroxy-4-(octyloxy)benzophenone, 2-Hydroxy-4-n-octoxybenzophenone, 2-Hydroxy-4-octoxybenzophenone, 4-(Octyloxy)-2-hydroxybenzophenone, 4-n-Octyloxy-2-hydroxybenzophenone, ADK Stab 1413, ARO 8, Adeka 1413, Aduvex 248, Advastab 46, Anti-UV P, Asahi 1413, Benzon OO, Benzophenone 12, C 81, Carstab 700, Chimassorb 81, Cyasorb 531, Cyasorb UV 531, Cyasorb UV 532, Cytec UV 531, GW 531, Hostavin ARO 8, LA 1413, Lowilite 22, Mark 1413, NSC 163400, Octabenzone, Ongrostab HOB, Rhodialux P, Sanduvor 3035, Seesorb 102, Seikalizer E, Specta-Sorb UV 531, Spectra-Sorb UV 531, Sumisorb 130, UF 4, UV 1, UV 1 (ultraviolet absorber), UV 531, Uvinul 3008, Uvinul M 408, V 130, Viosorb 130, Zislizer E
IUPAC Name (2-hydroxy-4-octoxyphenyl)-phenylmethanone
SMILES CCCCCCCCOc1ccc(C(=O)c2ccccc2)c(O)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C21H26O3/c1-2-3-4-5-6-10-15-24-18-13-14-19(20(22)16-18)21(23)17-11-8-7-9-12-17/h7-9,11-14,16,22H,2-6,10,15H2,1H3
Formula Weight 326.1882
Chemical Name or Material 2-Hydroxy-4-(octyloxy)benzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.