missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 2-hydroxyisobutyryl-Histone H1.2 (Lys63) Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5144296

Product Code. 17998962

  • € 401.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

2-hydroxyisobutyryl-Histone H1.2 (Lys63) Polyclonal Antibody for ICC/IF, ELISA

Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H1 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2-hydroxyisobutyryl-Histone H1.2 (Lys63)
Polyclonal
Unconjugated
H1-2
0610008C09Rik; CTL-1; H1 histone family, member 2; H1 VAR.1; H1.2; H1.2 linker histone, cluster member; H1-2; H1C; H1f2; H1s-1; H1var1; His1a; Hist1h1c; histone 1, H1c; histone cluster 1, H1c; histone H1; histone H1.1; histone H1.2; Histone H1c; histone H1d; Histone H1s-1; LOC684681; MGC3992; similar to Histone H1.2 (H1 VAR.1) (H1c)
Rabbit
Antigen affinity chromatography
RUO
3006
-20°C or -80°C if preferred
Liquid
ELISA, Immunocytochemistry
0.08 mg/mL
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
P16403
H1-2, HIST1H1C
Peptide sequence around site of 2-hydroxyisobutyryl-Lys (63) derived from Human Histone H1.2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.