missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylphenyl isocyanate, 95%, Thermo Scientific™

Catalog Number p-4323262
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
56309-56-9
C10H11NO
161.21
MWBSQVPLYJZPTG-UHFFFAOYSA-N
2-isopropylphenyl isocyanate, 1-isocyanato-2-isopropylbenzene, o-isopropylphenyl isocyanate, 2-isopropylphenylisocyanate, 1-isocyanato-2-propan-2-yl benzene, 1-isocyanato-2-isopropyl-benzene, 2-methylethyl benzenisocyanate, acmc-1avxp, isopropylphenyl isocyanate
91797
1-isocyanato-2-propan-2-ylbenzene
CC(C)C1=CC=CC=C1N=C=O
This item is not returnable. View return policy
56309-56-9
C10H11NO
161.21
MWBSQVPLYJZPTG-UHFFFAOYSA-N
2-isopropylphenyl isocyanate, 1-isocyanato-2-isopropylbenzene, o-isopropylphenyl isocyanate, 2-isopropylphenylisocyanate, 1-isocyanato-2-propan-2-yl benzene, 1-isocyanato-2-isopropyl-benzene, 2-methylethyl benzenisocyanate, acmc-1avxp, isopropylphenyl isocyanate
91797
1-isocyanato-2-propan-2-ylbenzene
CC(C)C1=CC=CC=C1N=C=O
CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 1.0190g/mL
Boiling Point 66.0°C to 67.0°C (5.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.019
Formula Weight 161.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Isopropylphenyl isocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Isopropylphenyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'