Learn More
2-Methoxy-3-pyridinecarboxaldehyde, 96%, Thermo Scientific™
Catalog Number p-4229281
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
71255-09-9
C7H7NO2
137.14
PIFFMIDNNWOQLK-UHFFFAOYSA-N
2-methoxynicotinaldehyde, 2-methoxy-3-pyridinecarboxaldehyde, 2-methoxy-pyridine-3-carbaldehyde, 2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-methoxy, 2-methoxy-3-pyridine carboxaldehyde, 3-pyridinecarboxaldehyde,2-methoxy, 2-methoxy-3-pyridinecarbaldehyde, pubchem15124, 2-methoxy-3-formyl-pyridine
12533391
2-methoxypyridine-3-carbaldehyde
COC1=C(C=CC=N1)C=O
71255-09-9
C7H7NO2
137.14
PIFFMIDNNWOQLK-UHFFFAOYSA-N
2-methoxynicotinaldehyde, 2-methoxy-3-pyridinecarboxaldehyde, 2-methoxy-pyridine-3-carbaldehyde, 2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-methoxy, 2-methoxy-3-pyridine carboxaldehyde, 3-pyridinecarboxaldehyde,2-methoxy, 2-methoxy-3-pyridinecarbaldehyde, pubchem15124, 2-methoxy-3-formyl-pyridine
12533391
2-methoxypyridine-3-carbaldehyde
COC1=C(C=CC=N1)C=O
CAS Min % 95.0
CAS Max % 100.0
Melting Point 25°C
Color Colorless to Yellow
Boiling Point 200°C to 201°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 137.14
Percent Purity 96%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Methoxy-3-pyridinecarboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methoxy-3-pyridinecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.