missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-pyridinecarboxylic acid, 97%, ACROS Organics™

Brand:  Acros Organics 442410050

Additional Details : CAS : 105596-63-2 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11996431

  • € 185.00 / 5g


Estimated Shipment: 27-07-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

105596-63-2
153.14
2-methoxyisonicotinic acid, 2-methoxy-4-pyridinecarboxylic acid, 2-methoxy-isonicotinic acid, 4-pyridinecarboxylic acid, 2-methoxy, 2-methoxy-isonicotinicacid, 2-methoxy-4-picolinic acid, pubchem9293, ksc179m4j, 4-pyridinecarboxylicacid, 2-methoxy, 2-methyloxy-4-pyridinecarboxylic acid
2-methoxypyridine-4-carboxylic acid
C7H7NO3
DPNDWFVORIGXQO-UHFFFAOYSA-N
819942
COC1=NC=CC(=C1)C(=O)O

Specifications

2-Methoxy-4-pyridinecarboxylic acid
Brown to White
5g
96.0
96% min. (HPLC)
2-methoxyisonicotinic acid, 2-methoxy-4-pyridinecarboxylic acid, 2-methoxy-isonicotinic acid, 4-pyridinecarboxylic acid, 2-methoxy, 2-methoxy-isonicotinicacid, 2-methoxy-4-picolinic acid, pubchem9293, ksc179m4j, 4-pyridinecarboxylicacid, 2-methoxy, 2-methyloxy-4-pyridinecarboxylic acid
COC1=NC=CC(=C1)C(=O)O
153.14
153.14
97%
Authentic
186°C to 188°C
105596-63-2
100.0
C7H7NO3
DPNDWFVORIGXQO-UHFFFAOYSA-N
2-methoxypyridine-4-carboxylic acid
819942
Solid or Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Methoxy-4-pyridinecarboxylic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates