Learn More
2-Methoxy-5-(trifluoromethoxy)aniline, 97%, Thermo Scientific™
Product Code. 15494907
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494907 1 g 1g
15404917 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15494907 Supplier Thermo Scientific Alfa Aesar Supplier No. H32219.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 660848-57-7
Molecular Formula C8H8F3NO2
Molecular Weight (g/mol) 207.152
MDL Number MFCD09832305
InChI Key MKAPTOAKEPIGOA-UHFFFAOYSA-N
Synonym 2-methoxy-5-trifluoromethoxy aniline, 2-methoxy-5-trifluoromethoxy phenylamine
PubChem CID 22506981
IUPAC Name 2-methoxy-5-(trifluoromethoxy)aniline
SMILES COC1=C(C=C(C=C1)OC(F)(F)F)N
Specifications
Refractive Index 1.4762
Quantity 1 g
UN Number UN2810
Formula Weight 207.15
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-(trifluoromethoxy)aniline