missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 11411280
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11411280 1 g 1g
11421280 5 g 5g
11431280 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11411280

Brand: Thermo Scientific Alfa Aesar H26362.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as an intermediate in synthetic chemistry.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Methoxy-5-(trifluoromethyl)benzoic acid is used as an intermediate in synthetic chemistry.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, bases and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4864-01-1
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.15
MDL Number MFCD01862091
InChI Key NAKZCKOHULJEID-UHFFFAOYSA-N
PubChem CID 13585909
IUPAC Name 2-methoxy-5-(trifluoromethyl)benzoic acid
SMILES COC1=C(C=C(C=C1)C(F)(F)F)C(=O)O

Specifications

Melting Point 108°C to 111°C
Quantity 1 g
Synonym 2-methoxy-5-trifluoromethyl benzoic acid, benzoic acid, 2-methoxy-5-trifluoromethyl, pubchem10415, acmc-209wn0, benzoic acid,2-methoxy-5-trifluoromethyl, 2-methyloxy-5-trifluoromethyl benzoic acid, 2-methoxy-5-trifluoromethyl benzoicacid
Solubility Information Insoluble in water.
Formula Weight 220.15
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.