Learn More
2-Methoxy-5-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
Product Code. 11461210
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461210 1 g 1g
11471210 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11461210 Supplier Thermo Scientific Alfa Aesar Supplier No. H26329.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Mostly existing as pharmaceutical and agrochemical products.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 685126-88-9
Molecular Formula C9H8BrF3O
Molecular Weight (g/mol) 269.061
MDL Number MFCD06660318
InChI Key ZGKOXCPOSPRGFQ-UHFFFAOYSA-N
Synonym 2-methoxy-5-trifluoromethyl benzyl bromide, 2-bromomethyl-1-methoxy-4-trifluoromethyl benzene, 2-methoxy-5-trifluoromethyl benzylbromide, 2-bromomethyl-1-methoxy4-trifluoromethyl-benzene
PubChem CID 17750810
IUPAC Name 2-(bromomethyl)-1-methoxy-4-(trifluoromethyl)benzene
SMILES COC1=C(C=C(C=C1)C(F)(F)F)CBr
Specifications
Melting Point 34°C to 37°C
Quantity 1 g
UN Number UN3261
Solubility Information Insoluble in water.
Formula Weight 269.06
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)benzyl bromide