Learn More
2-Methoxy-6-ethylnapthalene, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Ethyl-6-methoxynaphthalene (Ethylnerolin) |
| CAS | 21388-17-0 |
| Molecular Formula | C13 H14 O |
| Quantity | 100 mg |
| Synonym | 2-Ethyl-6-methoxynaphthalene, Naproxen Imp. J (EP), Ethylnerolin |
| InChI Formula | InChI=1S/C13H14O/c1-3-10-4-5-12-9-13(14-2)7-6-11(12)8-10/h4-9H,3H2,1-2H3 |
| SMILES | CCc1ccc2cc(OC)ccc2c1 |
| IUPAC Name | 2-ethyl-6-methoxynaphthalene |
| Molecular Weight (g/mol) | 186.25 |
| Formula Weight | 186.1045 |