Learn More
2-Methoxy-6-(methylamino)pyridine, 98%, Thermo Scientific™
Product Code. 11344466
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11344466
Brand: Thermo Scientific Alfa Aesar L20048.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 88569-83-9
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD00191557
InChI Key GXHQLRBOZRJHPZ-UHFFFAOYSA-N
Synonym 2-methoxy-6-methylamino pyridine, 2-methoxy-6-methylaminopyridine, 6-methoxy 2-pyridyl methylamine, 6-methoxy-n-methyl-pyridin-2-amine, pubchem15374, acmc-1bifn, 6-methoxy-2-methylaminopyridine, methoxymethylaminopyridine, 6-methoxy-n-methyl-2-pyridinamine
PubChem CID 7019168
IUPAC Name 6-methoxy-N-methylpyridin-2-amine
SMILES CNC1=CC=CC(OC)=N1
Specifications
Refractive Index 1.5595
Quantity 5 g
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 2-Methoxy-6-(methylamino)pyridine