Learn More
2-Methyl-1-[4-(methylsulfonyl)phenyl]-1-propanone, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-1-[4-(methylsulfonyl)phenyl]-1-propanone |
| CAS | 53207-59-3 |
| Molecular Formula | C11H14O3S |
| Quantity | 25 mg |
| Synonym | p-Methylsulfonylisobutyrophenone, 2-Methyl-1-[4-(methylsulfonyl)phenyl]-1-propanone |
| InChI Formula | InChI=1S/C11H14O3S/c1-8(2)11(12)9-4-6-10(7-5-9)15(3,13)14/h4-8H,1-3H3 |
| SMILES | CC(C)C(=O)c1ccc(cc1)S(=O)(=O)C |
| IUPAC Name | 2-methyl-1-(4-methylsulfonylphenyl)propan-1-one |
| Molecular Weight (g/mol) | 226.29 |
| Formula Weight | 226.0664 |