missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals

Catalog Number 15426186
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
58960-00-2
C9H11NS
165.254
MFCD12027245
QZSCUJZRZIIVBM-UHFFFAOYSA-N
2h-1,4-benzothiazine, 3,4-dihydro-2-methyl, 3,4-dihydro-2-methyl-2h-1,4-benzothiazine, 2-methyl-3,4-dihydro-2h-benzo b 1,4 thiazine
13183653
2-methyl-3,4-dihydro-2H-1,4-benzothiazine
CC1CNC2=CC=CC=C2S1
This item is not returnable. View return policy
58960-00-2
C9H11NS
165.254
MFCD12027245
QZSCUJZRZIIVBM-UHFFFAOYSA-N
2h-1,4-benzothiazine, 3,4-dihydro-2-methyl, 3,4-dihydro-2-methyl-2h-1,4-benzothiazine, 2-methyl-3,4-dihydro-2h-benzo b 1,4 thiazine
13183653
2-methyl-3,4-dihydro-2H-1,4-benzothiazine
CC1CNC2=CC=CC=C2S1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 165.25
Percent Purity 97%
Chemical Name or Material 2-Methyl-3,4-dihydro-2H-1,4-benzothiazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'