Learn More
2-Methyl-4-trimethylsilyl-1-buten-3-yne, 97%
Product Code. 11319688
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319688 10 g 10g
11329688 2 g 2g
11309688 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11319688 Supplier Thermo Scientific Alfa Aesar Supplier No. 044189.09
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
CAS: 18387-60-5 | C8H14Si | 138.285 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18387-60-5
Molecular Formula C8H14Si
Molecular Weight (g/mol) 138.285
MDL Number MFCD00190206
InChI Key HRGBALJHGYAWBL-UHFFFAOYSA-N
Synonym 2-methyl-4-trimethylsilyl-1-buten-3-yne, 2-methyl-4-trimethylsilylbut-1-en-3-yne, acmc-1cdn8, trimethylsilylisopropenylacetylene, trimethyl 3-methylbut-3-en-1-ynyl silane, 3-methyl-3-butene-1-ynyl trimethylsilane, trimethyl 3-methyl-3-buten-1-ynyl silane #, trimethyl 3-methylbut-3-en-1-yn-1-yl silane, silane,trimethyl 3-methyl-3-buten-1-yn-1-yl, 2-methyl-4-trimethylsilyl-1-buten-3-yne,
PubChem CID 579646
IUPAC Name trimethyl(3-methylbut-3-en-1-ynyl)silane
SMILES CC(=C)C#C[Si](C)(C)C
Specifications
Density 0.778g/mL
Boiling Point 32°C (15 mmHg)
Quantity 10 g
UN Number UN1993
Formula Weight 138.28
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-4-trimethylsilyl-1-buten-3-yne